SC 150-S7 (230V/50Hz) HEATING BATH CIRCULATOR STEEL, SC150-S7, Amb+13 TO +150 C, 4-7 L BATH VOLUME, 2.0 kW HEATER, PUMP 17 L/MIN & 300 MBAR USB UL/CSA, 1 * 1 items
Supplier: THERMO ELECTRON LED
SC 150-S7 (230V/50Hz) HEATING BATH CIRCULATOR STEEL, SC150-S7, Amb+13 TO +150 C, 4-7 L BATH VOLUME, 2.0 kW HEATER, PUMP 17 L/MIN & 300 MBAR USB UL/CSA, 1 * 1 items
Learn more About VWR
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on +353 1 88 22222.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on +353 1 88 22222.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at eurega_services@eu.vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organisation. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
Product(s) marked with this symbol are discontinued - sold till end of stock. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service on +353 1 8822222.