Syringe cleaner
Supplier: HAMILTON BONADUZ
76615 76610
HAMI76615EA 1690 EUR
HAMI76615 HAMI76610
Syringe cleaner
Sterilisers Loop Sterilisers
Specifically designed for rapidly cleaning 7000 series modified Microliter™ syringes, this syringe cleaner also effectively cleans Microliter™ and Gastight® syringes. A combination of heat (approximately 370 °C) and vacuum vaporises and extracts contaminants from the needle. The syringe cleaner is ineffective for septa-plugged needles, seised plungers and other irreparable needles.
Generally, Microliter™ or Gastight® syringes require a lower cleaning temperature than the Modified Microliter™ syringes.
Caution: Do not heat the syringes with cemented needles above 50 °C. ThermalMore Product Information