Reversed phase HPLC columns, PRP-h5
Supplier: HAMILTON BONADUZ
79261 79262 79272 79273 79263 79274 79267 79268 79277 79278 79270 79271 79296 79921
554-1392EA 982 EUR
554-1392 554-1393 554-1399 554-1400 554-1394 554-1401 554-1395 554-1396 554-1402 554-1403 554-1397 554-1398 HAMI79296 HAMI79921
Reversed phase HPLC columns, PRP-h5
Chromatography Columns
PRP-h5 polimeric reversed phase columns are ideal for applications that require a high degree of column-to-column and batch to batch consistency (quality assurance, clinical trials). Unlike C8 or C18 columns, PRP-h5 has no stationary phase coating. The integral reversed phase characteristics of the PRP-h5 column eliminate the need for special coating techniques. Since there is no stationary phase to hydrolyse, the column maintains its performance characteristics longer than many C8 or C18 columns. PRP-h5 columns are packed with a poly(styrene-divinylbenzene) material.
Order Now
SPECIFICATIONS
Environmentally Preferable
Max. pressure 5000 psi
pH range 1 - 13
Temperature limit 5 - 85 °C