Crown stopwatch, ABS
Supplier: HANHART
111.0117-00
HANH111.0117-00EA 309 EUR
HANH111.0117-00
Crown stopwatch, ABS
Timers Analogue Stopwatches
Analogue stopwatch with functional ABS plastic case and pendant ring.
  • Very lightweight and easy to handle
  • Pin lever, 1 jewel
  • 1/5 seconds, 30 minutes