VWR® EML 200 Test Sieve Shakers
Supplier: VWR Collection
510-2043EA 2530 EUR
510-2043 510-2044 510-2046 510-2047 510-2037 510-2038 510-2049 510-2050 510-2041 510-2040 HAVE209882186 HAVE209882209 HAVE209882216 HAVE209882193 HAVE209882094
VWR® EML 200 Test Sieve Shakers
Shakers Sieve Shakers
The EML 200 series offers different types of units. All models are suitable for 50 to 203 mm Ø sieves, a maximum sample/batch load of 3 kg, and have a cast aluminium housing and improved clamping system.
  • Digital timer: 0 to 99 minutes or constant operation
  • All shakers have a cover with inspection glass which allows samples to be viewed during sieving
EML 200 Pure units are ideal for standard analysis:
Feature two given regulated amplitudes to select from: Fine or coarse
Operation in set intervals of 10 seconds
Classic 'eco' clamping system
Order Now
SPECIFICATIONS
Environmentally Preferable