Magnetic Stirrers, Hei-PLATE Mix 'n' Heat Ultimate
Supplier: Heidolph Instruments GmbH & Co.KG
506-41100-03 506-42100-03 506-41150-00
HEID506-41100-03EA 2220 EUR
HEID506-41100-03 HEID506-42100-03 HEID506-41150-00
Magnetic Stirrers, Hei-PLATE Mix 'n' Heat Ultimate
Stirrers Magnetic Stirrers
Hei-PLATE Mix 'n' Heat Ultimate is equipped with numerous interfaces for automated laboratory processes. The RS232 and USB B ports allow a control via our integrated Hei-PROCESS solution.
The USB A interface at the front allows data logging via a USB stick. For more comfort and safety, you can connect the magnetic stirrer with the electric Heidolph lab jack via the USB C interface. There are three heating modes for temperature control: FAST for a quick heating, as well as PRECISE and PRECISE+ for the exact control of your target temperature without overshoot - this is how sensitiveMore Product Information