Culture media, in tubes
Supplier: MERCK LIFE SCIENCE
1463030100
HEIP1463030100EA 209 EUR
HEIP1463030100
Culture media, in tubes
Media for Microbiology Prepared Media for Microbiology
Sabouraud dextrose is recommended for the microbial enumeration tests.