Ultra low temperature freezers, with 4 inner doors, –40 °C, TDE series
Supplier: Thermo Scientific
TDE40040LV TDE40040FV TDE50040FV TDE60040FV
HERATDE40040LVEA 13990 EUR
HERATDE40040LV HERATDE40040FV HERATDE50040FV HERATDE60040FV
Ultra low temperature freezers, with 4 inner doors, –40 °C, TDE series
Freezers Ultra Low Temperature Freezers
Thermo Scientific™ TDE Series ‒40 °C ultra-low temperature freezers, powered by H-drive, feature three upright models, maximizing storage capacity from 40000 up to 60000 2 ml vials. The TDE Series is a sustainable and reliable solution with ultimate sample security and operational savings for every laboratory.
  • Powered by H-drive, save up to 25% in energy usage and heat rejection over conventional refrigerant ultra-low freezers
  • Easy-to-use set-up wizard, event log, temperature mapping and data downloads
  • Capacitive touch even when using latex gloves
  • Touch-
More Product Information