Capillary columns, HP-PLOT Q
Supplier: Agilent
19091P-QO3 19091P-QO4 19091P-QO4E 19095P-QO3 19095P-QO4 19095P-QO4E 19095P-QO3E 19091P-QO3LTM 19091P-QO3PT 19091P-QO4LTM 19091P-QO4PT 19095P-QO3LTM 19095P-QO3PT 19095P-QO4LTM 19095P-QO4PT
HEWL19091P-QO3EA 1070 EUR
HEWL19091P-QO3 HEWL19091P-QO4 HEWL19091P-QO4E HEWL19095P-QO3 HEWL19095P-QO4 HEWL19095P-QO4E HEWL19095P-QO3E HEWL19091P-QO3LTM HEWL19091P-QO3PT HEWL19091P-QO4LTM HEWL19091P-QO4PT HEWL19095P-QO3LTM HEWL19095P-QO3PT HEWL19095P-QO4LTM HEWL19095P-QO4PT
Capillary columns, HP-PLOT Q
Chromatography Columns
General purpose columns for solvents, sulfur compounds, oxygenated compounds, C1 to C3 isomers, methane, air/CO, and alkanes upto C12. Ideal for ethane, ethylene and ethyne separation.
  • Improved resolution than conventional packed columns
  • Minimal conditioning time of an hour
Bonded polystyrene-divinylbenzene
Max. temperature: 270/290 °C
Suitable replacement for gas-solid columns.