ISIS supplies for 7500
Supplier: Agilent
5064-8016 5064-8017 5064-8018 5064-8026 5064-8021 5064-8022 5064-8020 G3138-65121 5064-8027 5064-8019 5064-8029 5064-8032 5064-8033 G3138-65117 G3138-65118 G3138-65119 G3138-65122 G3138-65125 G3138-65158 5182-2581
HEWL5064-8016EA 249 EUR
HEWL5064-8016 HEWL5064-8017 HEWL5064-8018 HEWL5064-8026 HEWL5064-8021 HEWL5064-8022 HEWL5064-8020 HEWLG3138-65121 HEWL5064-8027 HEWL5064-8019 HEWL5064-8029 HEWL5064-8032 HEWL5064-8033 HEWLG3138-65117 HEWLG3138-65118 HEWLG3138-65119 HEWLG3138-65122 HEWLG3138-65125 HEWLG3138-65158 HEWL5182-2581
ISIS supplies for 7500
Chromatography Supplies Chromatography Spare Parts
High quality supplies and components for the Integrated Sample Introduction System (ISIS) for use with the 7500 ICP-MS range.
  • ETFE sample tubing with a 0,8 mm Int.Ø and 1,6 mm Ext.Ø is also available along with PTFE sample tubing with a 2 mm Int.Ø and 3 mm Int.Ø
  • Alternatively, PFA sample tubing with 0,3 mm Int.Ø and 1,6 mm Ext.Ø can be provided
  • A variety of joints are available in tee, cross or union form
  • PTFE nuts and front and back ferrules for either 3 mm or 1,6 mm Ext.Ø can be provided
  • Spiral tubing, peristaltic pump tubing adapters and tubing clamps,
More Product Information