Fused silica tubing cutter
Supplier: Agilent
5183-4620 420-1000
HEWL5183-4620EA 924 EUR
HEWL5183-4620 HEWL420-1000
Fused silica tubing cutter
Cutters
Cutting tool, for fused silica tubing, with rotating diamond blade.