HPLC columns, ZORBAX® 80 Å StableBond®
Supplier: Agilent
7995218-344 7995218-585 7995218-595 830990-902 831975-902 831975-932 833975-902 833975-932 835975-902 835975-905 835975-906 835975-912 846975-902 860975-902 860975-905 860975-912 863600-902 863953-902 863953-905 863953-906 863954-302 863954-305 863954-306 863954-312 865600-902 866953-902 871700-902 873700-902 873700-932 875700-902 875700-932 880975-202 880975-205 880975-212 880975-302 880975-305 880975-306 880975-309 880975-312 880975-902 880975-905 880975-906 880975-912 880975-914 883700-905 883700-912 883700-922 883975-302 883975-305 883975-306 883975-309 883975-312 883975-902 883975-905 883975-906 883975-909 883975-912 830990-914 835975-914 846975-914 860975-914 861753-914 861953-914 863953-914 866953-914 871700-914 883975-914 846975-202 861953-902 861953-905 861954-306 883975-202 861954-302 861954-305 822975-902 823975-902 821975-902 822700-902 823700-902 821700-902 821700-932 821975-932 822700-932 822975-932 823700-932 823975-932 825700-902 825700-932 825975-902 825975-932 861753-902 861753-912 861953-912 861954-309 861954-312 861954-314 863600-905 863954-309 863954-314 866735-902 880975-314 883975-314 884950-567 884975-202 827700-902 824700-902 828700-902 827975-902 828975-906 820700-902 826700-902 820700-906 827700-905 827975-905 827700-912 827975-912 828975-902 824700-906 827700-906 827700-914 820700-912 827975-306 829975-306 828700-906 820700-905 824700-905 824700-912 824700-914 824975-302 824975-305 824975-306 824975-902 824975-905 824975-906 824975-912 824975-914 826700-906 826975-302 826975-306 826975-902 826975-906 827975-302 827975-305 827975-312 827975-314 827975-906 827975-914 828700-905 828700-912 828700-914 828975-302 828975-305 828975-306 828975-312 828975-314 828975-905 828975-912 828975-914 829975-302 829975-305 829975-312 829975-902 829975-905 829975-906 829975-912 829975-914 832975-902 833975-912 834975-902 874700-902 830975-906 858700-914 857700-305 858700-905 859700-902 858700-902 857700-905 857700-902 857700-914 858700-314 823750-902 866953-302 859768-901 821725-902 820750-902 827975-902K 827975-912K 827975-914K 828975-902K 828975-912K 828975-914K 829975-902K 829975-912K 829975-914K 835975-902K 835975-912K 835975-914K 857700-302K 858700-302K 859700-302K 859700-902K 861953-902K 861953-912K 861953-914K 863953-912K 863953-914K 880975-902K 880975-912K 880975-914K 883975-902K 883975-906K 883975-912K 883975-914K 884950-567K 863953-902K
HEWL7995218-344EA 1250 EUR
HEWL7995218-344 HEWL7995218-585 HEWL7995218-595 HEWL830990-902 HEWL831975-902 HEWL831975-932 HEWL833975-902 HEWL833975-932 HEWL835975-902 HEWL835975-905 HEWL835975-906 HEWL835975-912 HEWL846975-902 HEWL860975-902 HEWL860975-905 HEWL860975-912 HEWL863600-902 554-1285 HEWL863953-905 554-1286 HEWL863954-302 HEWL863954-305 HEWL863954-306 HEWL863954-312 HEWL865600-902 HEWL866953-902 HEWL871700-902 HEWL873700-902 HEWL873700-932 HEWL875700-902 HEWL875700-932 HEWL880975-202 HEWL880975-205 HEWL880975-212 HEWL880975-302 HEWL880975-305 HEWL880975-306 HEWL880975-309 HEWL880975-312 554-1289 HEWL880975-905 554-1290 554-1291 HEWL880975-914 HEWL883700-905 HEWL883700-912 554-1294 HEWL883975-302 HEWL883975-305 HEWL883975-306 HEWL883975-309 HEWL883975-312 554-1296 HEWL883975-905 554-1297 HEWL883975-909 HEWL883975-912 HEWL830990-914 HEWL835975-914 HEWL846975-914 HEWL860975-914 HEWL861753-914 HEWL861953-914 HEWL863953-914 HEWL866953-914 HEWL871700-914 HEWL883975-914 HEWL846975-202 HEWL861953-902 HEWL861953-905 HEWL861954-306 HEWL883975-202 HEWL861954-302 HEWL861954-305 HEWL822975-902 HEWL823975-902 HEWL821975-902 HEWL822700-902 HEWL823700-902 HEWL821700-902 HEWL821700-932 HEWL821975-932 HEWL822700-932 HEWL822975-932 HEWL823700-932 HEWL823975-932 HEWL825700-902 HEWL825700-932 HEWL825975-902 HEWL825975-932 HEWL861753-902 HEWL861753-912 HEWL861953-912 HEWL861954-309 HEWL861954-312 HEWL861954-314 HEWL863600-905 HEWL863954-309 HEWL863954-314 HEWL866735-902 HEWL880975-314 HEWL883975-314 HEWL884950-567 HEWL884975-202 HEWL827700-902 HEWL824700-902 HEWL828700-902 554-1417 HEWL828975-906 HEWL820700-902 HEWL826700-902 HEWL820700-906 HEWL827700-905 HEWL827975-905 554-1416 HEWL827975-912 554-1418 HEWL824700-906 HEWL827700-906 HEWL827700-914 554-1426 554-1427 554-1428 HEWL828700-906 HEWL820700-905 HEWL824700-905 HEWL824700-912 HEWL824700-914 HEWL824975-302 HEWL824975-305 HEWL824975-306 HEWL824975-902 HEWL824975-905 HEWL824975-906 HEWL824975-912 HEWL824975-914 HEWL826700-906 HEWL826975-302 HEWL826975-306 HEWL826975-902 HEWL826975-906 HEWL827975-302 HEWL827975-305 HEWL827975-312 HEWL827975-314 HEWL827975-906 HEWL827975-914 HEWL828700-905 HEWL828700-912 HEWL828700-914 HEWL828975-302 HEWL828975-305 HEWL828975-306 HEWL828975-312 HEWL828975-314 HEWL828975-905 HEWL828975-912 HEWL828975-914 HEWL829975-302 HEWL829975-305 HEWL829975-312 HEWL829975-902 HEWL829975-905 HEWL829975-906 HEWL829975-912 HEWL829975-914 HEWL832975-902 HEWL833975-912 HEWL834975-902 HEWL874700-902 HEWL830975-906 HEWL858700-914 HEWL857700-305 HEWL858700-905 HEWL859700-902 HEWL858700-902 HEWL857700-905 HEWL857700-902 HEWL857700-914 HEWL858700-314 HEWL823750-902 HEWL866953-302 HEWL859768-901 HEWL821725-902 HEWL820750-902 HEWL827975-902K HEWL827975-912K HEWL827975-914K HEWL828975-902K HEWL828975-912K HEWL828975-914K HEWL829975-902K HEWL829975-912K HEWL829975-914K HEWL835975-902K HEWL835975-912K HEWL835975-914K HEWL857700-302K HEWL858700-302K HEWL859700-302K HEWL859700-902K HEWL861953-902K HEWL861953-912K HEWL861953-914K HEWL863953-912K HEWL863953-914K HEWL880975-902K HEWL880975-912K HEWL880975-914K HEWL883975-902K HEWL883975-906K HEWL883975-912K HEWL883975-914K HEWL884950-567K HEWL863953-902K
HPLC columns, ZORBAX® 80 Å StableBond®
Chromatography Columns
ZORBAX® 80 Å StableBond® columns are made using bulky, patented, unique silanes that sterically protect the siloxane bond. Acid labile endcapping reagents are not used.
  • Longest column lifetime and best reproducibility for low pH separations - down to pH 1
  • Patented stable column chemistry allows use at high temperature and low pH without degradation
  • High purity (type B) silica for good peak shape