Soyabean Casein Digest Medium

Supplier: HIMEDIA

LQ193CC-5X200M
HIMELQ193CC-5X200MEA 136 EUR
HIMELQ193CC-5X200M
Soyabean Casein Digest Medium
Media for Microbiology Dehydrated Media for Microbiology

Sterility test medium also used for cultivation of soyabean casein digest medium.


  • Colour: Light yellow coloured opalascent solution
  • Appearance: Sterile soyabean casein digest medium with 0,5% Soya Lecithin and 4% Tween 20 in glass bottle
  • Quantity of medium: 200 ml of medium in glass bottle
  • Cultural response: Cultural characteristics observed after an incubation as specified


Soyabean casein digest medium with 0,5% Soya Lecithin and 4% Tween 20 is used for the detection of microorganisms on surfaces of sanitary importance. Tryptone and soya peptone makes this medium nutritious by providing nitrogeneous and carbonaceous compounds, long chain peptides, vitamins and other essential nutrients for the growth of microorganisms.


Caution: Read the label before opening the container. Wear protective gloves/protective clothing/eye protection/face protection. Follow good microbiological lab practices while handling specimens and culture. Standard precautions as per established guidelines should be followed while handling specimens.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR