Stoppers for standard ground joints, glass
Supplier: HIRSCHMANN
8900352 8900360 8900365 8900370 8900375 8900380 8900386 8900390 8900393 8902370 8902352 8902360 8902365 8902375 8902380 8902386 8902390 8902393
8900352.EA 5.2 EUR
8900352. 8900360. 8900365. 8900370. 8900375. 8900380. 8900386. 8900390. 8900393. 8902370. HIRS8902352 HIRS8902360 HIRS8902365 HIRS8902375 HIRS8902380 HIRS8902386 HIRS8902390 HIRS8902393
Stoppers for standard ground joints, glass
Stoppers Glass Stoppers
Stoppers from clear or amber glass.
Certifications: DIN 12252
Order Now
SPECIFICATIONS
Environmentally Preferable