HPLC/UHPLC columns, Kromasil® Classic, 100 Å
Supplier: HICHROM
MH3NHA15 MH2CLA10 MH3CMA10 M05CLJ15 MH3CMC25 M05CMC25 M10WLQ25 M10WLA25 M10WLP25
HISMMH3NHA15EA 752 EUR
HISMMH3NHA15 HISMMH2CLA10 HISMMH3CMA10 HISMM05CLJ15 HISMMH3CMC25 HISMM05CMC25 HISMM10WLQ25 HISMM10WLA25 HISMM10WLP25
HPLC/UHPLC columns, Kromasil® Classic, 100 Å
Chromatography Columns
The Kromasil® Classic 100 Å family of products is used to separate and purify molecules of up to 10000 Da. Drug candidates for the pharmaceutical, natural products and API industries are separated and purified using Kromasil® Classic 100 Å columns.
  • Spherical porous high purity silica
  • Analytical to preparative particle silica: 1,8; 2,5; 3,5; 5; 10; 13 and 16 µm
  • 1,8 µm columns suitable for UHPLC conditions; 2,5 µm columns suitable for HPLC and UHPLC conditions
  • Unbonded (SIL) and derivatised (C1, C4, C8, C18, C18(w), NH2 and Phenyl) phases available
  • C18(w) is
More Product Information