Trichloroacetic acid (TCA) 99-100.5% (dried basis), meets analytical specification of Ph. Eur., USP 21, Fluka™

Supplier: Honeywell Chemicals
Danger

Synonyms: TCA

27242H-1KG 27242H-500G
HONC27242H-1KGEA 166 EUR
HONC27242H-1KG HONC27242H-500G
Trichloroacetic acid (TCA) 99-100.5% (dried basis), meets analytical specification of Ph. Eur., USP 21, Fluka™
Trichloroacetic acid (TCA)
Formula: Cl₃CCOOH
Boiling Pt: 196 °C
Density: 1,63 g/cm3
Flash Pt: 113 °C
MDL Number: MFCD00004177
CAS Number: 76-03-9
UN: 1839
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ae34130684af3f73eccad44eac686e325855232d&vid=b600bf2ba67ce275263e6172675e131d3f263837&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (calculated to the dried substance) 99 - 100.5 %
Loss on drying (on silica gel, 18h) Max. 0.5 %
Sulfated ash Max. 0.02 %
Iron (Fe) Max. 0.002 %
Chloride (Cl) Max. 0.005 %
Sulfate (SO₄) Max. 0.05 %
Appearance of the solution complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR