Pyridine, Puriss. p.a. ACS, Riedel-de Haën™

Supplier: Honeywell Chemicals
Danger

Synonyms: Azabenzene, Azine

33501B-200L
HONC33501B-200LEA 6710 EUR
HONC33501B-200L
Pyridine, Puriss. p.a. ACS, Riedel-de Haën™
Pyridine
Formula: C₅H₅N
MW: 79.1 g/mol
Boiling Pt: 115 °C (1,013 hPa)
Melting Pt: –42 °C
Density: 0.98 g/cm3 (20 °C)
Flash Pt: 20 °C
Storage Temperature: Ambient
MDL Number: MFCD00011732
CAS Number: 110-86-1
EINECS: 203-809-9
UN: 1282
ADR: 3 II
Merck Index: 13,08060

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR