Butyl acetate ≥99.5%, Puriss. p.a. ACS, Riedel-de Haën™

Supplier: Honeywell Chemicals
Danger

45860-2.5L 45860-1L 45860-35KG
45860-2.5LEA 209 EUR
45860-2.5L 45860-1L HONC45860-35KG
Butyl acetate ≥99.5%, Puriss. p.a. ACS, Riedel-de Haën™
Butyl acetate

ACS GENERAL USE SOLVENTS


  • Value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • Suitable for less demanding applications such as routine isocratic HPLC
  • Variations and decrease issues traceable to solvent contamination
  • All solvents are tested to meet ACS general use specifications


Beilstein Number: 1741921
RTECS Number: AF7350000
UNSPSC Code: 12191602

Formula: CH₃COO(CH₂)₃CH₃
Boiling Pt: 126,2 °C (1.013 hPa)
Melting Pt: -78 °C
Density: 0,881 g/cm³ (20 °C)
Flash Pt: 10,4 %(V)
Storage Temperature: Ambient
MDL Number: MFCD00009445
CAS Number: 123-86-4
EINECS: 204-658-1
UN: 1123
ADR: 3,III
Merck Index: 13,01534
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=403329de943f8cb2ea6cd9d73ee35957d2140aa7&vid=682ab82de177b2985e94089408f7efebde59926d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Autoignition Temperature 790 °F
concentration 90 - 110 ng/µl
purity (GC) Min. 97 %
refractive index (n 20/D) 1.328 - 1.331
Non-volatile matter Max. 0.001 %
Free acid (as CH₃COOH) Max. 0.01 %
Aluminium (Al) Max. 0.5 ppm
Barium (Ba) Max. 0.1 ppm
Bismuth (Bi) Max. 0.1 ppm
Boron (B) Max. 0.02 ppm
Calcium (Ca) Max. 0.5 ppm
Cadmium (Cd) Max. 0.05 ppm
Cobalt (Co) Max. 0.02 ppm
Chromium (Cr) Max. 0.02 ppm
Copper (Cu) Max. 0.02 ppm
Iron (Fe) Max. 0.1 ppm
Potassium (K) Max. 0.5 ppm
Lithium (Li) Max. 0.1 ppm
Magnesium (Mg) Max. 0.1 ppm
Manganese (Mn) Max. 0.02 ppm
Molybdenum (Mo) Max. 0.1 ppm
Sodium (Na) Max. 0.5 ppm
Nickel (Ni) Max. 0.02 ppm
Lead (Pb) Max. 0.1 ppm
Tin (Sn) Max. 0.1 ppm
Strontium (Sr) Max. 0.1 ppm
Zinc (Zn) Max. 0.1 ppm
n-Butyl formate Max. 0.1 %
1-Butanol (GC) Max. 0.2 %
n-Butyl propionate Max. 0.1 %
Reaction against H₂SO₄ complying
APHA Max. 10


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR