Acetonitrile Reag. Ph. Eur., gradient grade for HPLC, Burdick & Jackson™

Supplier: Honeywell Chemicals
Danger

65708-2.5L 65708-200L
HONC65708-2.5LEA 409 EUR
HONC65708-2.5L HONC65708-200L
Acetonitrile Reag. Ph. Eur., gradient grade for HPLC, Burdick & Jackson™
Acetonitrile
Formula: H₃CCN
Boiling Pt: 82 °C (1,013 hPa)
Melting Pt: –46 °C
Density: 0,7857 g/cm3 (20 °C)
Flash Pt: 6 °C
Storage Temperature: Ambient
MDL Number: MFCD00001878
CAS Number: 75-05-8
EINECS: 200-835-2
UN: 1648
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cef8be50fcca82050c5bda827ad9c5f57f4c5e27&vid=b80b5c39e654fcf6904b18b58e22a816e8e138b7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance complying
Assay (GC) Min. 99.9 %
Boiling range 80-82 °C Min. 95 %
Density (D 20/20) 0.782 - 0.784
Refractive index (n 20/D) 1.3435 - 1.3445
Non-volatile matter Max. 0.0002 %
Water (Karl Fischer) Max. 0.01 %
Acidity Max. 0.0002 meq/g
Alkalinity Max. 0.0001 meq/g
Transmittance at 195 nm Min. 75 %
Transmittance at 200 nm Min. 93 %
Transmittance from 230 nm Min. 99 %
HPLC Gradient at 210 nm Max. 3 mAU
HPLC Gradient at 254 nm Max. 0.5 mAU
Baseline drift at 210 nm Max. 15 mAU
Fluorescence (chinin) at 254 nm Max. 1 ppb
Fluorescence (chinin) at 365 nm Max. 0.5 ppb
Phthalates as dibutylphthalate Max. 0.1 mg/l
Reaction of the 10 % solution to Litmus complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR