Petroleum spirit, 40-60°C for gas chromatography, Burdick & Jackson™

Supplier: Honeywell Chemicals

65748-200L-RC
HONC65748-200L-RCEA 3150 EUR
HONC65748-200L-RC
Petroleum spirit, 40-60°C for gas chromatography, Burdick & Jackson™
Petroleum spirit
Boiling Pt: 40…60 °C
Flash Pt: –30 °C
MDL Number: MFCD00081849
CAS Number: 64742-49-0
UN: 1268
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR