Acetonitrile ≥99.8% (by GC) for preparative HPLC, Riedel-de Haën™

Supplier: Honeywell Chemicals
Danger

82063-200L 82063-395L 82063-18L
HONC82063-200LEA 25440 EUR
HONC82063-200L HONC82063-395L HONC82063-18L
Acetonitrile ≥99.8% (by GC) for preparative HPLC, Riedel-de Haën™
Acetonitrile
Formula: H₃CCN
Boiling Pt: 82 °C (1,013 hPa)
Melting Pt: –46 °C
Density: 0,7857 g/cm3 (20 °C)
Flash Pt: 6 °C
Storage Temperature: Ambient
MDL Number: MFCD00001878
CAS Number: 75-05-8
EINECS: 200-835-2
UN: 1648
ADR: 3,II
REACH: 01-2119471307-38
Merck Index: 13,00071
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cef8be50fcca82050c5bda827ad9c5f57f4c5e27&vid=b80b5c39e654fcf6904b18b58e22a816e8e138b7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) Min. 99.8 %
Non-volatile matter Max. 0.0001 %
Water (Karl Fischer) Max. 0.1 %
Free alkali (as NH₃) Max. 0.0005 %
Transmittance at 220 nm Min. 50 %
Transmittance from 240 nm Min. 99 %
Explosion Limit 16 %
Autoignition Temperature 973 °F
Vapor Pressure 72.8 mmHg ( 20 °C)

SPECIFICATIONS

Environmentally Preferable Safer Human and Environmental Health
Environmental certification Chem21 Solvent Selection Guide recommended by default and after discussion

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR