Holmium ICP & ICP-MS standards, (CRMs), Fluka™
Supplier: Honeywell Chemicals
E1392-100ML
HONCE1392-100MLEA 94.1 EUR
HONCE1392-100ML
Holmium ICP & ICP-MS standards, (CRMs), Fluka™
Standards Cationic Standards Holmium Standards
Certified Reference Materials (CRMs)
  • Traceable to SRM from NIST
Certifications: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).