Calibration mix majors for AA and ICP-OES
Supplier: Honeywell Chemicals
E2022-100ML
HONCE2022-100MLEA 383 EUR
HONCE2022-100ML
Calibration mix majors for AA and ICP-OES
Standards Cationic Standards Multi-Element Standards
Certified Reference Material (CRM).
500 mg/L each of Ca, Fe, K, Mg, Na in HNO₃ 5%