Combined Seven Anion Standard I for IC
Supplier: Honeywell Chemicals
E2029-100ML
HONCE2029-100MLEA 243.36 EUR
HONCE2029-100ML
Combined Seven Anion Standard I for IC
Standards Anionic Standards Multi-Element Anioinic Standards
Certified Reference Material (CRM).
150 mg/L of PO43-, SO42-; 100 mg/L of NO2-, Br-, NO3-; 30 mg/L Cl-; 20 mg/L F- in H₂O