VIS wavecal solution for ICP
Supplier: Honeywell Chemicals
E2122-100ML
HONCE2122-100MLEA 191 EUR
HONCE2122-100ML
VIS wavecal solution for ICP
Standards Cationic Standards Multi-Element Standards
Certified Reference Material (CRM).
50 mg/L K, 10 mg/L each of La, Li, Mn, Na, Sr, 1 mg/L each of Ba, Ca in HNO₃ 2%