Nickel (II) nitrate 10 g/L in nitric acid 1% matrix modifier solution, for atomic absorption spectroscopy, Fluka™

Supplier: Honeywell Chemicals

E4605-50ML
HONCE4605-50MLEA 311.72 EUR
HONCE4605-50ML
Nickel (II) nitrate 10 g/L in nitric acid 1% matrix modifier solution, for atomic absorption spectroscopy, Fluka™
Nickel (II) nitrate

Certified Reference Material, 10 g/L Ni(NO₃)₂ in 1% HNO₃


Certifications: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).

Formula: N₂NiO₆
Density: 1,01 g/cm3
Storage Temperature: Ambient
MDL Number: MFCD00011139
CAS Number: 13138-45-9

Order Now

Specification Test Results

Matrix HNO₃ 1%
Concentration Ni(NO₃)₂ 10,000 mg/L
Quality ISO/IEC17025:2017(ANAB),ISO17034:2016(ANAB)
Form Solution
Density 1.01g/cm3


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR