Quinalphos Analytical standard, reference material, Fluka™

Supplier: Honeywell Chemicals

R1801-100MG
HONCR1801-100MGEA 43.7 EUR
HONCR1801-100MG
Quinalphos Analytical standard, reference material, Fluka™
Standards Organic Standards

Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials  and organic reference materials.


  • Produced according to ISO 9001
  • Suitable for a wide range of industries and applications
  • Detailed Certificate of Analysis included with every delivery

Order Now

SPECIFICATIONS

Description Quinalphos
Environmentally Preferable
Physical form Crystalline
CAS No. 13593-03-8
MDL No. MFCD00078719
Molecular weight 298.30 g/mol
Melting point 31 - 32 °C
UN No. 2811
Storage conditions 2-8°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR