Gram's iodine solution, Sigma-Aldrich®

Supplier: SIGMA ALDRICH MICROSCOPY

Synonyms: Lugol's iodine, Iodine and potassium iodide, Potassium triiodide

MQ500

HT90232-1L HT902-8FOZ
HT90232-1LEA 186 EUR
HT90232-1L HT902-8FOZ
Gram's iodine solution, Sigma-Aldrich®
Gram's iodine solution

Used as a kit component in Gram Stain Procedure (Sigma Procedure HT90). Iodine, 0.33%, and potassium iodide, 0.66%.

Formula: I₃K
MW: 419.812 g/mol
Storage Temperature: Room Temperature
MDL Number: MFCD00134128
CAS Number: 12298-68-9
IVDLogo

Order Now

Specification Test Results

Appearance (Colour) Orange-Brown
Appearance (Form) Solution
Appearance (Turbidity)
Essentially free of particles/precipitate
Clear
Performance Conforms
Gram positive organisms are purple-black in both bacteriology smears and tissue sections; Gram negative organisms are red in both bacteriology smears and tissue sections
Expiration Date Period 24 months


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR