HyClone™ Donor Equine Serum, U.S. origin, HyClone products (Cytiva)
Supplier: HyClone products (Cytiva)
SH30074.04 SH30074.04IR25
HYCLSH30074.04EA 163 EUR
HYCLSH30074.04 SH30074.04IR25
HyClone™ Donor Equine Serum, U.S. origin, HyClone products (Cytiva)
Sera (Serum)
The animals are fed an enriched diet to ensure proper nutrition and are fasted prior to bleeding to reduce lipid concentration. The horses are bled in an enclosed facility that is specially equipped for aseptic collection. Coggins tests for equine infectious anemia are routinely performed on each horse.
  • Tested for equine infectious anemia
  • Complete traceability back to original source
  • Virus panel testing according to 9 CFR 113,53
onor Equine Serum is filtered through three sequential 100 nm (0,1 μm) pore size-rated filters. Before dispensing, each lotMore Product Information