Affinity chromatography column, MAbPac™ Protein A
Supplier: Thermo Fisher Scientific
082539
HYPE082539EA 1331.63 EUR
HYPE082539
Affinity chromatography column, MAbPac™ Protein A
Chromatography Columns
For fast, accurate titer analysis of monoclonal antibodies (MAbs) in harvest cell cultures. This column provides high throughput and accurate analyses through a combination of low backpressure and high efficiency. Its unique format makes automation of loading, binding, elution and collection using biocompatible systems quick and easy. A non-porous, polymeric resin with a hydrophobic divinylbenzene core and a hydrophilic surface optimised for affinity separation makes this column rugged and reproducible.
  • Complete cycle time, including equilibration, of two minutes
  • Linear over a wide
More Product Information