SPME fibers and arrows
Supplier: Thermo Fisher Scientific
36SP01P1 36SP01P3 36SP03P3 36SP05T1 36SP05T3 36SP06E1 36SP06E3 36SP08A1 36SP08A3 36SP08M5 36SP09C1 36SP09C3 36SP10P1 36SP10P3
HYPE36SP01P1EA 420 EUR
HYPE36SP01P1 HYPE36SP01P3 HYPE36SP03P3 HYPE36SP05T1 HYPE36SP05T3 HYPE36SP06E1 HYPE36SP06E3 HYPE36SP08A1 HYPE36SP08A3 HYPE36SP08M5 HYPE36SP09C1 HYPE36SP09C3 HYPE36SP10P1 HYPE36SP10P3
SPME fibers and arrows
Chromatography Supplies Chromatography Spare Parts
Solid Phase Micro Extraction (SPME) uses a thin-fused silica fiber coated with a sorbent which enables extraction of compounds of interest from liquid or gaseous samples through absorption or adsorption. The fiber is then moved to the injector and the compounds thermally desorbed. The SPME consumables come in two formats SPME fibers and SPME arrows.
All SPME fibers have a standard length of 10 mm and a 23 gauge needle.
SPME arrows have a phase length of 20 mm but vary in diameter. A specific SPME arrow tool on the autosampler is required.
Ordering information: For method development,More Product Information