Glass snap top vials, 2 ml, level 3 high performance applications, SureSTART™
Supplier: Thermo Fisher Scientific
6PRV11-1P 6PRV11-1PF 6PRV11-1PT
HYPE6PRV11-1PEA 25.7 EUR
HYPE6PRV11-1P HYPE6PRV11-1PF HYPE6PRV11-1PT
Glass snap top vials, 2 ml, level 3 high performance applications, SureSTART™
Vials
Choose Thermo Scientific™ SureSTART™ 2 ml glass snap top vials, performance level 3, for high performance applications, when sensitivity is a must, and when you are running many samples.
Our level 3 vials are our highest performance vials, designed to protect your most valuable samples and deliver consistent, reproducible results for your most demanding analyses. These 2 ml clear glass snap vials are specification certified to ensure their physical dimensions and autosampler compatibility. Use 11 mm snap caps or 11 mm crimp caps to seal this vial. Our snap caps provide a very tight sealMore Product Information