N,O-Bis(trimethylsilyl)trifluoroacetamide with 1% trimethylchlorosilane

Supplier: Thermo Fisher Scientific
Danger

TS-38834
HYPETS-38834EA 0 EUR
HYPETS-38834
N,O-Bis(trimethylsilyl)trifluoroacetamide with 1% trimethylchlorosilane
N,O-Bis(trimethylsilyl)trifluoroacetamide with 1% trimethylchlorosilane
Formula: C₈H₁₈F₃NOSi₂
MDL Number: MFCD00008269
CAS Number: 25561-30-2
EINECS: 247-103-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR