Immersion Circulators, ICC 150 lite pro 12
Supplier: IKA
10013212
IKAA10013212EA 2300 EUR
IKAA10013212
Immersion Circulators, ICC 150 lite pro 12
Circulating Thermostats Open Circulator Systems Thermostat Controllers
The compact immersion circulators of the lite series are powerful and of high quality, with a focus on essential basic functions, excellent performance and efficient operation.
  • Max. temperature in stainless steel bath (pro): 150 °C
ICC 150 lite is suitable for tempering liquids up to 150 °C. The integrated pressure/suction pump allows ICC 150 lite to be used for internal and external applications.
To make configuration of your temperature control system as convenient as possible, IKA offers 'ready-to-connect' packages with all the necessaryMore Product Information