Recirculating chiller, environmentally friendly, RC 2 lite
Supplier: IKA
25007940
IKAA25007940EA 4300 EUR
IKAA25007940
Recirculating chiller, environmentally friendly, RC 2 lite
Circulating Thermostats Closed Circulator Systems Cooling Circulators
Compact recirculating chiller with powerful 400 W cooling capacity, designed for simple cooling tasks down to –10 °C. The RC 2 lite is the perfect peripheral device for the cooling of rotary evaporators, reflux condensers or as a cooling source for devices that dissipate heat with the help of a heat exchanger (double-walled grinding containers, cooled incubators, laboratory reactors, cooling coils). The easily accessible and easy-to-clean mesh filter along with low filling volume of just one litre enables fast cooling. Thanks to the natural refrigerant R290, the RC 2 lite is remarkablyMore Product Information