Dry vacuum and chemical resistant diaphragm pumps (MPC)
Supplier: Welch by Gardner Denver
412522 412543 412422-02 412443-02 412521 412781 412782 412783 412983 412422-10 412443-10 412421-02 412021 415722 412922-06
ILMV412522EA 1420 EUR
ILMV412522 181-0515 ILMV412422-02 ILMV412443-02 ILMV412521 ILMV412781 ILMV412782 ILMV412783 ILMV412983 ILMV412422-10 ILMV412443-10 ILMV412421-02 ILMV412021 ILMV415722 ILMV412922-06
Dry vacuum and chemical resistant diaphragm pumps (MPC)
Pumps Vacuum Diaphragm Pumps
The MPC chemical duty diaphragm pumps use PTFE and other fluorinated plastics for the wetted parts to allow aggressive solvent and acid vapours to be pumped. If the pump should handle harsh chemical vapours, then the superior chemical resistant X2 version with higher grade seals and thicker coatings are the best choice. These are well known for quiet operation and running smoothness.
  • Different formats are available (1, 2, 4 head diaphragm pumps)
  • Low weight and compact design
  • Can be moved to different locations based on applications
They are availableMore Product Information