Anti-IgG Guinea Pig Polyclonal Antibody

Supplier: ImmunoReagents
GP-003-CBIO
IMMRGP-003-CBIOEA 103 EUR
IMMRGP-003-CBIO
Anti-IgG Guinea Pig Polyclonal Antibody
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Guinea Pig
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR