Anti-IgG Donkey Polyclonal Antibody (DyLight® 650)

Supplier: ImmunoReagents
IR1272
IMMRIR1272EA 135 EUR
IMMRIR1272
Anti-IgG Donkey Polyclonal Antibody (DyLight® 650)
Antibodies
The ImmunoReagents' DyLight® 650 conjugates provide vibrant far-red fluorescence with comparable or improved performance compared to other dyes for fluorescent applications. DyLight® 650 dye conjugates have tight excitation/emission spectra that are ideal for multi-Colour applications. DyLight® 650 conjugated secondary antibodies are available for use in fluorescence microscopy, flow cytometry, Western blotting, ELISA, high-content screening and other array platforms. Excitation/Emission = 652 nm/672 nm Emission Colour = Far Red (Similar Dyes: Alexa Fluor 647, Cy5).

  • Origin: US

Order Now

SPECIFICATIONS

Antigen symbol IgG
Antigen name Immunoglobulin G
Environmentally Preferable
Antibody type Secondary
Clonality Polyclonal
Conjugation DyLight® 650
Reactivity Rat
Host Donkey
Isotype IgG
Western blot Yes
ELISA Yes
ImmunoChemistry Yes
ImmunoPrecipitation Yes
ImmunoFluorescence Yes
Flow cytometry Yes
Immunogen Rat IgG
Format Purified IgG
Form Lyophilised
Purification Affinity purified using solid phaseRat IgG
Concentration 1 mg/ml (E 1% at 280 nm = 13,0)
Cross adsorption No
Storage buffer 10 mM Sodium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 1% (w/v) BSA, Protease/IgG free
Storage temperature 2...8 °C
Shipping temperature 2...8 °C
Tested applications Fluoresence ELISA, Fluorescence-Activated Cell Sorting

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR