Anti-IgG Donkey Polyclonal Antibody (AP (Alkaline Phosphatase))

Supplier: ImmunoReagents
IR1292
IMMRIR1292EA 186 EUR
IMMRIR1292
Anti-IgG Donkey Polyclonal Antibody (AP (Alkaline Phosphatase))
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Donkey
Isotype: IgG
Reactivity: Sheep
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR