Anti- Donkey Polyclonal Antibody (Unconjugated)

Supplier: ImmunoReagents
IR2412
IMMRIR2412EA 103 EUR
IMMRIR2412
Anti- Donkey Polyclonal Antibody (Unconjugated)
Antibodies
Type:
Antigen:
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Donkey
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR