Dithiothreitol (DTT, Cleland's reagent) ≥99.5%, Ultrapure for molecular biology

Supplier: Thermo Fisher Scientific
Warning

J15397.22 J15397.14 J15397.06
J15397.22EA 1460 EUR
J15397.22 J15397.14 J15397.06
Dithiothreitol (DTT, Cleland's reagent) ≥99.5%, Ultrapure for molecular biology
Dithiothreitol (DTT, Cleland's reagent)

Used as a protective agent for SH groups.

Formula: C₄H₁₀O₂S₂
Boiling Pt: 125…130 °C (12 mmHg)
Melting Pt: 40…43 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Freezer
MDL Number: MFCD00004877
CAS Number: 3483-12-3
EINECS: 222-468-7
Merck Index: 14,03376
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5022472c3f3baef75a703791c548fc6504df810f&vid=81529732db015cb66c1f106092309ae5714325b7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (SH) ≥99.5%
Assay Absorbance (260 nm, 0.1 M, 1 cm) ≤0.40
Assay Absorbance (280 nm, 0.1 M, 1 cm) ≤0.08
DNase Passes Test
Impurity Content, OxidiSed DTT ≤0.3%
Protease Activity Passes Test
RNase Passes Test

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR