Mineral oil, white (petroleum) light, high purity

Supplier: VWR Chemicals
Warning

Synonyms: Matrix oil, Mineral oil, Paraffin oil

J217-100MLEA 68.1 EUR
J217-100ML J217-500ML
Mineral oil, white (petroleum) light, high purity
White mineral oil (petroleum)
Boiling Pt: 300 °C (1013 hPa)
Density: 0,872 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00131611
CAS Number: 8042-47-5
EINECS: 232-455-8

Order Now

Specification Test Results

Specific Gravity 0.838 – 0.854
Viscosity @ 40 °C 14.2 – 17 cS


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR