Okadaic acid ≥98%

Supplier: Thermo Fisher Scientific
Danger

J60155.XS J60155.XG J60155.IX
J60155.XSEA 506 EUR
J60155.XS J60155.XG J60155.IX
Okadaic acid ≥98%
Okadaic acid
Formula: C₄₄H₆₈O₁₃
Storage Temperature: Freezer
MDL Number: MFCD00083455
CAS Number: 78111-17-8
UN: 3462
ADR: 6.1,I
Merck Index: 14,6819

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR