Aminophylline, anhydrous 98%, powder

Supplier: Thermo Fisher Scientific
Danger

J60705.14 J60705.22
J60705.14EA 51.6 EUR
J60705.14 J60705.22
Aminophylline, anhydrous 98%, powder
Aminophylline

A non-selective phosphodiesterase inhibitor


Aminophylline exhibits vasodilatory and anti-inflammatory activity and is occasionally clinically used to treat chronic obstructive pulmonary disorder (COPD).


In animal models of allergen-induced lung inflammation, aminophylline decreases levels of IL-5, IL-8, and eosinophils. It is mostly used to prepare solutions for intravenous injections.

Formula: C₇H₈N₄O₂
Melting Pt: 269…274 °C
Storage Temperature: Freezer
MDL Number: MFCD00013221
CAS Number: 317-34-0
EINECS: 206-264-5
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Purity 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR