Laemmli SDS sample buffer, reducing (6X)

Supplier: Thermo Fisher Scientific

J61337.ACEA 104 EUR
J61337.AC J61337.AD
Laemmli SDS sample buffer, reducing (6X)
Buffers Electrophoresis Buffers Loading Buffers

Physical Form: Liquid
Appearance: Blue
Applications: For protein sample preparation to be used in the Laemmli SDS-PAGE system

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR