H-7 dihydrochloride

Supplier: Thermo Fisher Scientific

Synonyms: 5-((2-Methylpiperazin-1-yl)sulphonyl)isoquinoline dihydrochloride, Protein kinase inhibitor H-7 2HCl

J61697.MA J61697.MF
J61697.MAEA 185 EUR
J61697.MA J61697.MF
H-7 dihydrochloride
H-7 dihydrochloride

Protein kinase inhibitor. Inhibits cyclic nucleotide dependent protein kinase (PKA) and protein kinase C.


Powder

Formula: C₁₄H₁₇N₃O₂S·2HCl
MW: 364.29 g/mol
Melting Pt: 191…193 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00036961
CAS Number: 108930-17-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR