LDS-sample buffer (4X), non-reducing
Supplier: Thermo Fisher Scientific
J61894.ACEA 86.5 EUR
J61894.AC J61894.AD
LDS-sample buffer (4X), non-reducing
Buffers Electrophoresis Buffers Loading Buffers
LDS-sample buffer (4X), non-reducing is used in SDS-polyacrylamide gel electrophoresis (SDS-PAGE). It is used in denaturing gels and is compatible with both coomassie and silver staining and Western blotting applications.
Physical Form: Liquid
Appearance: Blue
This buffer contains: 988 mM Tris-HCl, 2.04 mM EDTA, 8% LDS, 40% glycerol, 0.88% Coomassie blue and 0.7 mM phenol red at pH 8.5.
Order Now
SPECIFICATIONS
Environmentally Preferable
Appearance Blue
Physical form Liquid