Lansoprazole ≥98%

Supplier: Thermo Fisher Scientific
Warning

J62008.ME J62008.03 J62008.06
J62008.MEEA 85.8 EUR
J62008.ME J62008.03 J62008.06
Lansoprazole ≥98%
Lansoprazole

Purity: 98+% CAS No.: 103577-45-3 Molecular Formula: C16H14F3N3O2S Formula Weight: 369.36 Storage Temperature: 2°C to 8°C Physical Form: Crystalline powder Melting Point: 173-175° Merck Reference: 14,5362 MDL No.: MFCD00866873


A proton pump inhibitor

Formula: C₁₆H₁₄F₃N₃O₂S
Melting Pt: 173…175 °C
MDL Number: MFCD00866873
CAS Number: 103577-45-3
Merck Index: 14,5362

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR