Neutral red

Supplier: Thermo Fisher Scientific

J63054.03 J63054.06 J63054.14
J63054.03EA 40.9 EUR
J63054.03 J63054.06 J63054.14
Neutral red
Neutral red

CAS No.: 553-24-2 Molecular Formula: C15H17ClN4 Formula Weight: 288.78 Physical Form: Powder Appearance: Dark green to black Melting Point: 290° dec. Beilstein Reference: 3918740 Merck Reference: 14,6488 MDL No.: MFCD00012651


Nuclear counterstain useful in pH range 6.8 - 8.0

Formula: C₁₅H₁₇ClN₄
Melting Pt: 290 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012651
CAS Number: 553-24-2
EINECS: 209-035-8
Merck Index: 14,6488

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR