β-Cyclodextrine

Supplier: Thermo Fisher Scientific

J63161.14 J63161.22
J63161.14EA 43.3 EUR
J63161.14 J63161.22
β-Cyclodextrine
β-Cyclodextrine

Use to solubilize non-polar compounds such a fatty acids, lipids and cholesterol. Reported useful for the selective precipitation of enantiomeric, positional or structural isomers


Use to solubilize non-polar compounds such a fatty acids, lipids and cholesterol. Reported useful for the selective precipitation of enantiomeric, positional or structural isomers

Formula: C₄₂H₇₀O₃₅
Melting Pt: 290 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00078139
CAS Number: 7585-39-9
EINECS: 231-493-2
Merck Index: 14,2718

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR